missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Fluorophenyl)-5-methylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64875.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15452269

  • £66.46 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Fluorophenyl)-5-methylbenzimidazole
226.25
MFCD18914842

Specifications

2-(3-Fluorophenyl)-5-methylbenzimidazole
Solid
226.25
1g
95%
MFCD18914842
Videos
SDS