missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Fluorophenyl)-5-methylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64875.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15462269

  • £331.78 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Fluorophenyl)-5-methylbenzimidazole
226.25
MFCD18914842

Specifications

2-(3-Fluorophenyl)-5-methylbenzimidazole
Solid
226.25
5g
95%
MFCD18914842
Videos
SDS