missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethylmaleic anhydride, 97%, Thermo Scientific Chemicals
29.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
2,3-Dimethylmaleic anhydride acts as a reagent in the preparation of maleimides. It is also employed as an amino group protecting agent for superoxide dismutase. Further, it serves as a reactant in the dissociation of ribosomal proteins. In addition to this, it is used in the preparation of dimethylmaleic acid dimethyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dimethylmaleic anhydride acts as a reagent in the preparation of maleimides. It is also employed as an amino group protecting agent for superoxide dismutase. Further, it serves as a reactant in the dissociation of ribosomal proteins. In addition to this, it is used in the preparation of dimethylmaleic acid dimethyl ester.
Solubility
Soluble in ethanol, ether and chloroform. Slightly soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
2,3-Dimethylmaleic anhydride acts as a reagent in the preparation of maleimides. It is also employed as an amino group protecting agent for superoxide dismutase. Further, it serves as a reactant in the dissociation of ribosomal proteins. In addition to this, it is used in the preparation of dimethylmaleic acid dimethyl ester.
Solubility
Soluble in ethanol, ether and chloroform. Slightly soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Melting Point | 93°C to 96°C |
Boiling Point | 222°C to 223°C |
Quantity | 5 g |
Beilstein | 112044 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethanol,ether and chloroform. Slightly soluble in water. |
Formula Weight | 126.11 |
Percent Purity | 97% |
Chemical Name or Material | 2,3-Dimethylmaleic anhydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'