missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 15481629
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5779-93-1
C9H10O
134.178
MFCD00798004
UIFVCPMLQXKEEU-UHFFFAOYSA-N
benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
34224
2,3-dimethylbenzaldehyde
CC1=CC=CC(=C1C)C=O
This item is not returnable. View return policy
5779-93-1
C9H10O
134.178
MFCD00798004
UIFVCPMLQXKEEU-UHFFFAOYSA-N
benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
34224
2,3-dimethylbenzaldehyde
CC1=CC=CC(=C1C)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.029 g/mL
Boiling Point 86°C to 88°C (10 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.553
Quantity 25 g
Beilstein 2238677
Formula Weight 134.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylbenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'