missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Dimethyl-6-nitroaniline, 98%

Brand:  Alfa Aesar™ A10412.18

Code : NEW

Additional Details : CAS Number : 59146-96-2 Weight : 0.05000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 15433027

  • £29.47 / 50g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2,3-Dimethyl-6-nitroaniline
C8H10N2O2
MFCD00010148
6-Nitro-2,3-xylidine
59146-96-2
166.18
UN2811

Specifications

2,3-Dimethyl-6-nitroaniline
98%
115° to 118°C
MFCD00010148
50g
59146-96-2
C8H10N2O2
166.18
UN2811
6-Nitro-2,3-xylidine
Videos
SDS