missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2-butanol, 98%, Thermo Scientific Chemicals

Catalog Number p-3757762
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
594-60-5
C6H14O
102.18
MFCD00004463
IKECULIHBUCAKR-UHFFFAOYSA-N
2,3-dimethyl-2-butanol, thexyl alcohol, isopropyldimethylcarbinol, 2-butanol, 2,3-dimethyl, acmc-1asco, 2,3-dimethyl-butan-2-ol, 2-butanol,2,3-dimethyl, ikeculihbucakr-uhfffaoysa, ch3 2chc oh ch3 2
11670
2,3-dimethylbutan-2-ol
CC(C)C(C)(C)O
This item is not returnable. View return policy
594-60-5
C6H14O
102.18
MFCD00004463
IKECULIHBUCAKR-UHFFFAOYSA-N
2,3-dimethyl-2-butanol, thexyl alcohol, isopropyldimethylcarbinol, 2-butanol, 2,3-dimethyl, acmc-1asco, 2,3-dimethyl-butan-2-ol, 2-butanol,2,3-dimethyl, ikeculihbucakr-uhfffaoysa, ch3 2chc oh ch3 2
11670
2,3-dimethylbutan-2-ol
CC(C)C(C)(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -14.0°C
Color Colorless
Density 0.8200g/mL
Boiling Point 120.0°C to 121.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2CHC(CH3)2OH
Refractive Index 1.4160 to 1.4180
Quantity 5 mL
Beilstein 01, 413
Specific Gravity 0.82
Formula Weight 102.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethyl-2-butanol
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethyl-2-butanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'