missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1H-indole-6-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3175078
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
103986-06-7
C11H11NO2
189.214
MFCD06799503
UDPOIMGOHQYJEE-UHFFFAOYSA-N
1h-indole-6-carboxylicacid, 2,3-dimethyl, 2,3-dimethylindole-6-carboxylic acid, acmc-1bpsb, 2,3-dimethyl-6-carboxyindole, 1h-indole-6-carboxylic acid, 2,3-dimethyl
2772885
2,3-dimethyl-1H-indole-6-carboxylic acid
CC1=C(NC2=C1C=CC(=C2)C(=O)O)C
This item is not returnable. View return policy
103986-06-7
C11H11NO2
189.214
MFCD06799503
UDPOIMGOHQYJEE-UHFFFAOYSA-N
1h-indole-6-carboxylicacid, 2,3-dimethyl, 2,3-dimethylindole-6-carboxylic acid, acmc-1bpsb, 2,3-dimethyl-6-carboxyindole, 1h-indole-6-carboxylic acid, 2,3-dimethyl
2772885
2,3-dimethyl-1H-indole-6-carboxylic acid
CC1=C(NC2=C1C=CC(=C2)C(=O)O)C
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethyl-1H-indole-6-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'