missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxyphenethylamine, 97%, Acros Organics

Brand:  Acros Organics 439710010

UNSPSC : 12161500

Code : 29

Additional Details : CAS Number : 3213-29-4 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 1b Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 11965311

  • £52.40 / 1g


EU Stock 4
Add to basket


Specifications

Chemical Identifiers

2, 3-Dimethoxyphenethylamine
97%
C10H15NO2
181.23

Specifications

2, 3-Dimethoxyphenethylamine
3213-29-4
97%
181.23
1g
Glass bottle
C10H15NO2
Safety and Handling

Safety and Handling

Product Identifier
  • 2,3-Dimethoxyphenethylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents