2,3-Dimethoxyphenethylamine, 97%, Acros Organics


Overview
Brand: Acros Organics

Manufacturer Part Number: 439710050

UNSPSC: 12161500

Code: 29

Additional Details:
Additional Details: CAS Number: 3213-29-4 Weight: 0.05500kg Transport: UN number : 2735 Chem class : 1b Pack group : III


Packaging Glass bottle
Quantity 5g

Product Code. 11975311

Quantity Price
1 £ 183.8 / 5g
EU Stock 3
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2, 3-Dimethoxyphenethylamine
Assay Percent Range 97%
Molecular Formula C10H15NO2
Formula Weight 181.23

Specification

Chemical Name or Material 2, 3-Dimethoxyphenethylamine
Quantity 5g
CAS 3213-29-4
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C10H15NO2
Formula Weight 181.23
Documentation

Technical documents