Learn More
Alfa Aesar™ 2,3-Dimethoxynaphthalene, 97%
Brand: Alfa Aesar™ H55523.03
Code : NEW
Additional Details : CAS Number : 10103-06-7 Weight : 0.00100kg
Quantity | 1g |
---|
Description
ApplicationsIt is used as a pharmaceutical intermediate.
Notes
Refrigerated (2-4°C). Keep away from oxidizing agents.
Chemical Identifiers
2,3-Dimethoxynaphthalene | |
10103-06-7 | |
188.23 | |
MFCD05664308 |
Specifications
2,3-Dimethoxynaphthalene | |
116° to 121°C | |
10103-06-7 | |
97% | |
MFCD05664308 |
White to yellow | |
1g | |
Powder | |
188.23 |