missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Dimethoxynaphthalene, 97%

Brand:  Alfa Aesar™ H55523.14

Code : NEW

Additional Details : CAS Number : 10103-06-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15421818

  • £422.80 / 25g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Description

Description

Applications
It is used as a pharmaceutical intermediate.

Notes
Refrigerated (2-4°C). Keep away from oxidizing agents.
Specifications

Chemical Identifiers

2,3-Dimethoxynaphthalene
10103-06-7
188.23
MFCD05664308

Specifications

2,3-Dimethoxynaphthalene
116° to 121°C
10103-06-7
97%
MFCD05664308
White to yellow
25g
Powder
188.23
Videos
SDS