missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzylamine, 99%, Thermo Scientific™

Catalog Number p-4228608
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4393-09-3
C9H13NO2
167.21
MFCD00052392
LVMPWFJVYMXSNY-UHFFFAOYSA-N
This item is not returnable. View return policy
4393-09-3
C9H13NO2
167.21
MFCD00052392
LVMPWFJVYMXSNY-UHFFFAOYSA-N
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 140.0°C to 142.0°C (15.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.11
Synonym 2,3-dimethoxybenzylamine, 2,3-dimethoxyphenyl methanamine, 1-2,3-dimethoxyphenyl methanamine, benzenemethanamine, 2,3-dimethoxy, 2,3-dimethoxy-benzylamine, 2,3-dimethoxy benzylamine, pubchem7399, 2,3-dimethoxybenzyl amine, acmc-2097fc
SMILES COC1=CC=CC(=C1OC)CN
IUPAC Name (2,3-dimethoxyphenyl)methanamine
PubChem CID 78106
Formula Weight 167.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'