missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Dimethoxy-beta-nitrostyrene, 98%

Brand:  Alfa Aesar™ A11347.06

Code : NEW

Additional Details : CAS Number : 37630-20-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11413774

  • £52.80 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2,3-Dimethoxy-beta-nitrostyrene
C10H11NO4
MFCD00068082
37630-20-9
209.19
1-(2,3-Dimethoxyphenyl)-2-nitroethylene

Specifications

2,3-Dimethoxy-beta-nitrostyrene
98%
80° to 82°C
MFCD00068082
1-(2,3-Dimethoxyphenyl)-2-nitroethylene
37630-20-9
C10H11NO4
209.19
5g
Videos
SDS