missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-112057
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


18.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
303-38-8
C7H6O4
154.12
MFCD00002446
GLDQAMYCGOIJDV-UHFFFAOYSA-N
pyrocatechuic acid, o-pyrocatechuic acid, 2-pyrocatechuic acid, 3-hydroxysalicylic acid, dobk, dhba, 2,3-dihydroxybenzoicacid, benzoic acid, 2,3-dihydroxy, catecholcarboxylic acid, 2,3-dihydroxy-benzoic acid
19
CHEBI:18026
2,3-dihydroxybenzoic acid
C1=CC(=C(C(=C1)O)O)C(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
303-38-8
C7H6O4
154.12
MFCD00002446
GLDQAMYCGOIJDV-UHFFFAOYSA-N
pyrocatechuic acid, o-pyrocatechuic acid, 2-pyrocatechuic acid, 3-hydroxysalicylic acid, dobk, dhba, 2,3-dihydroxybenzoicacid, benzoic acid, 2,3-dihydroxy, catecholcarboxylic acid, 2,3-dihydroxy-benzoic acid
19
CHEBI:18026
2,3-dihydroxybenzoic acid
C1=CC(=C(C(=C1)O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 204°C to 208°C
Color Beige to Brown or Gray-Pink
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CO2H
Quantity 5 g
Beilstein 10, 375
Formula Weight 154.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dihydroxybenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'