Learn More
Alfa Aesar™ 2,3-Dihydrobenzo[b]furan, 98%
Brand: Alfa Aesar™ A11368.14
Code : NEW
Additional Details : CAS Number : 496-16-2 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
2,3-Dihydrobenzo[b]furan | |
C8H8O | |
MFCD00005855 | |
14,2559 |
496-16-2 | |
120.15 | |
111928 | |
Coumaran; 2,3-Dihydro-2,3-benzofuran |
Specifications
2,3-Dihydrobenzo[b]furan | |
188° to 189°C | |
496-16-2 | |
C8H8O | |
MFCD00005855 | |
111928 | |
Coumaran; 2,3-Dihydro-2,3-benzofuran |
66°C (150°F) | |
25g | |
98% | |
120.15 | |
−21°C | |
14,2559 |