missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydrobenzo[b]furan-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 164.16 |
Percent Purity | 97% |
Chemical Name or Material | 2, 3-Dihydrobenzo[b]furan-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'