missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-2696921
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
115010-11-2
C8H7ClO3S
218.65
MFCD02677703
RVWYPBARHGPULM-UHFFFAOYSA-N
2,3-dihydrobenzofuran-5-sulfonyl chloride, 2,3-dihydro-1-benzofuran-5-sulfonoylchloride, 2,3-dihydro-1-benzofuran-5-sulfonoyl chloride, 2,3-dihydrobenzofuran-5-sulfonylchloride, 2,3-dihydro-benzofuran-5-sulfonyl chloride, 2,3-dihydrobenzo b furan-5-sulfonyl chloride, pubchem14874, 5-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzofuran-5-sulfonylchloride, chloro-2,3-dihydrobenzo b furan-5-ylsulfone
2776154
2,3-dihydro-1-benzofuran-5-sulfonyl chloride
ClS(=O)(=O)C1=CC2=C(OCC2)C=C1
This item is not returnable. View return policy
115010-11-2
C8H7ClO3S
218.65
MFCD02677703
RVWYPBARHGPULM-UHFFFAOYSA-N
2,3-dihydrobenzofuran-5-sulfonyl chloride, 2,3-dihydro-1-benzofuran-5-sulfonoylchloride, 2,3-dihydro-1-benzofuran-5-sulfonoyl chloride, 2,3-dihydrobenzofuran-5-sulfonylchloride, 2,3-dihydro-benzofuran-5-sulfonyl chloride, 2,3-dihydrobenzo b furan-5-sulfonyl chloride, pubchem14874, 5-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzofuran-5-sulfonylchloride, chloro-2,3-dihydrobenzo b furan-5-ylsulfone
2776154
2,3-dihydro-1-benzofuran-5-sulfonyl chloride
ClS(=O)(=O)C1=CC2=C(OCC2)C=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 218.66
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-5-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'