missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxine-6-sulfonyl chloride, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Especificaciones
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 234.66 |
Percent Purity | 97% |
Chemical Name or Material | 2, 3-Dihydro-1, 4-benzodioxine-6-sulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'