missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-5-carbothioamide, 95%, Maybridge

Brand:  Maybridge CC01418CB

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 690632-23-6 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10289242

  • £61.00 / 250mg


Estimated Shipment date
from Supplier 04-07-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide
94.5
95%
C9H9NO2S
MFCD05865104
690632-23-6
100.0
95%
195.24
2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide (IUPAC)

Specifications

195.24
690632-23-6
100.0
C9H9NO2S
Amber Glass Bottle
MFCD05865104
250mg
2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide
94.5
2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide (IUPAC)
95%
95%
Documentation

Technical Documents