Learn More
2,3-Dihydro-1,4-benzodioxine-5-carbothioamide, 95%, Maybridge
Brand: Maybridge CC01418DA
Code : 31
Additional Details : CAS Number : 690632-23-6 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : III
Quantity | 1g |
---|---|
Packaging | Amber Glass Bottle |
Chemical Identifiers
2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide | |
94.5 | |
95% | |
C9H9NO2S | |
MFCD05865104 |
690632-23-6 | |
100.0 | |
95% | |
195.24 | |
2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide (IUPAC) |
Specifications
2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide | |
690632-23-6 | |
100.0 | |
95% | |
C9H9NO2S | |
MFCD05865104 |
1g | |
94.5 | |
Amber Glass Bottle | |
95% | |
195.24 | |
2, 3-Dihydro-1, 4-benzodioxine-5-carbothioamide (IUPAC) |