missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4380086
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
851386-31-7
C6H3F2NO2
159.09
FFGHMEKFPCGYEG-UHFFFAOYSA-N
2,3-difluoroisonicotinic acid, 2,3-difluoro-4-pyridinecarboxylic acid, 2,3-difluoro-4-carboxypyridine, 4-pyridinecarboxylic acid, 2,3-difluoro, 2,3-fifluoroisonicotinic acid, 4-carboxy-2,3-difluoropyridine, 2,3-difluoroisonicotinicacid, pubchem18431, ksc447o0d, 5,6-difluoropyridine-4-carboxylic acid
11240637
2,3-difluoropyridine-4-carboxylic acid
C1=CN=C(C(=C1C(=O)O)F)F
This item is not returnable. View return policy
851386-31-7
C6H3F2NO2
159.09
FFGHMEKFPCGYEG-UHFFFAOYSA-N
2,3-difluoroisonicotinic acid, 2,3-difluoro-4-pyridinecarboxylic acid, 2,3-difluoro-4-carboxypyridine, 4-pyridinecarboxylic acid, 2,3-difluoro, 2,3-fifluoroisonicotinic acid, 4-carboxy-2,3-difluoropyridine, 2,3-difluoroisonicotinicacid, pubchem18431, ksc447o0d, 5,6-difluoropyridine-4-carboxylic acid
11240637
2,3-difluoropyridine-4-carboxylic acid
C1=CN=C(C(=C1C(=O)O)F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165°C to 167°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 159.09
Percent Purity 97%
Chemical Name or Material 2, 3-Difluoropyridine-4-carboxylic acid
compliance-icons
Product Identifier
  • 2,3-Difluoropyridine-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'