missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzylamine, 97%, ACROS Organics™

Brand:  Acros Organics 433370050

Code : 29

Additional Details : CAS Number : 72235-51-9 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 8 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10452970

  • £126.70 / 5g


Estimated Shipment
21-06-2018
Add to basket


Specifications

Chemical Identifiers

2, 3-Difluorobenzylamine
96.0
≥96.0%
C7H7F2N
MFCD00070793
72235-51-9
100.0
97%
143.14

Specifications

2, 3-Difluorobenzylamine
Clear colorless to pale yellow
Authentic
96.0
Glass bottle
≥96.0%
C7H7F2N
MFCD00070793
1.223g/mL
5g
72235-51-9
100.0
Liquid
97%
143.14
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,3-Difluorobenzylamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS