missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methoxybenzylamine, 97%, Thermo Scientific™

Catalog Number p-7029023
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
886500-75-0
C8H9F2NO
173.163
MFCD04115987
DBQHLJOOMYBKNQ-UHFFFAOYSA-N
2,3-difluoro-4-methoxybenzylamine, 2,3-difluoro-4-methoxyphenyl methanamine, 2,3-difluoro-4-methoxy-phenyl methanamine, 2,3-difluoro-4-methoxyphenyl methylamine, benzenemethanamine,2,3-difluoro-4-methoxy, 2,3-bis fluoranyl-4-methoxy-phenyl methanamine, 1-2,3-difluoro-4-methoxyphenyl methanamine
3817603
(2,3-difluoro-4-methoxyphenyl)methanamine
COC1=C(C(=C(C=C1)CN)F)F
This item is not returnable. View return policy
886500-75-0
C8H9F2NO
173.163
MFCD04115987
DBQHLJOOMYBKNQ-UHFFFAOYSA-N
2,3-difluoro-4-methoxybenzylamine, 2,3-difluoro-4-methoxyphenyl methanamine, 2,3-difluoro-4-methoxy-phenyl methanamine, 2,3-difluoro-4-methoxyphenyl methylamine, benzenemethanamine,2,3-difluoro-4-methoxy, 2,3-bis fluoranyl-4-methoxy-phenyl methanamine, 1-2,3-difluoro-4-methoxyphenyl methanamine
3817603
(2,3-difluoro-4-methoxyphenyl)methanamine
COC1=C(C(=C(C=C1)CN)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 32°C to 35°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 173.16
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-4-methoxybenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'