missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Difluoro-4-ethoxybenzeneboronic acid, 98%

Brand:  Alfa Aesar™ H64408.06

Code : NEW

Additional Details : CAS Number : 212386-71-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15481609

  • £85.30 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2,3-Difluoro-4-ethoxybenzeneboronic acid
201.96
19662857
212386-71-5
MFCD09258743
2,3-Difluoro-4-ethoxyphenylboronic acid

Specifications

2,3-Difluoro-4-ethoxybenzeneboronic acid
5g
Solid
201.96
19662857
151° to 154°C
212386-71-5
98%
MFCD09258743
2,3-Difluoro-4-ethoxyphenylboronic acid
Videos
SDS