missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxycytidine, 98%, ACROS Organics™

£76.14 - £316.40

Chemical Identifiers

CAS 7481-89-2
Molecular Formula C9H13N3O3
Molecular Weight (g/mol) 211.22
InChI Key WREGKURFCTUGRC-POYBYMJQSA-N
Synonym zalcitabine, 2',3'-dideoxycytidine, dideoxycytidine, ddcyd, hivid, ddc, cytidine, 2',3'-dideoxy, zalcitibine, ddc antiviral, 4-amino-1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one
PubChem CID 24066
ChEBI CHEBI:10101
IUPAC Name 4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1CC(OC1CO)N2C=CC(=NC2=O)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
13428668
SDS View Documents
Acros Organics
449501000
100mg Glass bottle
£76.14
100mg
View alternative products
Save Up to 
»
Estimated Shipment
20-05-2021
Please to purchase this item. Need a web account? Register with us today!
13438668
SDS View Documents
Acros Organics
449505000
500mg Glass bottle
£316.40
500mg
View alternative products
Save Up to 
»
Estimated Shipment
20-05-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7481-89-2
211.22
zalcitabine, 2',3'-dideoxycytidine, dideoxycytidine, ddcyd, hivid, ddc, cytidine, 2',3'-dideoxy, zalcitibine, ddc antiviral, 4-amino-1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one
CHEBI:10101
C1CC(OC1CO)N2C=CC(=NC2=O)N
C9H13N3O3
WREGKURFCTUGRC-POYBYMJQSA-N
24066
4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one

Specifications

2', 3'-Dideoxycytidine
216.0°C to 220.0°C
7481-89-2
C9H13N3O3
zalcitabine, 2',3'-dideoxycytidine, dideoxycytidine, ddcyd, hivid, ddc, cytidine, 2',3'-dideoxy, zalcitibine, ddc antiviral, 4-amino-1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one
WREGKURFCTUGRC-POYBYMJQSA-N
4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
24066
211.22
Glass bottle
100mg
97.5% min. (HPLC)
1510307
Solubility in water: soluble
C1CC(OC1CO)N2C=CC(=NC2=O)N
211.22
CHEBI:10101
98%
Safety and Handling

Safety and Handling

Product Identifier
  • 2',3'-Dideoxycytidine

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2

Hazard Statement
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS

RMF - Research Use or Further Mfg