missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyadenosine, 98%, Thermo Scientific Chemicals

Catalog Number p-7042952
Click to view available options
Quantity:
25 mg
50 mg
100 mg
This item is not returnable. View return policy
4097-22-7
C10H13N5O2
235.25
MFCD00010534
WVXRAFOPTSTNLL-NKWVEPMBSA-N
2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189
20039
[(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol
NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1
This item is not returnable. View return policy
4097-22-7
C10H13N5O2
235.25
MFCD00010534
WVXRAFOPTSTNLL-NKWVEPMBSA-N
2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189
20039
[(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol
NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 181°C to 184°C
Color White
Quantity 25 mg
Beilstein 619924
Merck Index 14,3101
Formula Weight 235.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2',3'-Dideoxyadenosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'