missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorothiophene, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | -37°C |
Color | Colorless to Yellow |
Density | 1.4700g/mL |
Boiling Point | 172°C to 174°C |
Flash Point | 78°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Refractive Index | 1.5687 |
Show More |
Safety and Handling
Product Identifier
- 2,3-Dichlorothiophene
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H301-Toxic if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'