missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorothiobenzamide, 97%, Thermo Scientific™

Catalog Number p-7029312
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
84863-83-2
C7H5Cl2NS
206.084
MFCD00221163
PRRMJTDPKZBCCQ-UHFFFAOYSA-N
2,3-dichlorothiobenzamide, 2,3-dichloro-thiobenzamide, 2,3-dichlorobenzothioamide, 2,3-dichlorobenzene-1-carbothioamide, amino 2,3-dichlorophenyl methane-1-thione, dichlorothiobenzamide, pubchem22984, acmc-20aoh4, benzenecarbothioamide, 2,3-dichloro
2799029
2,3-dichlorobenzenecarbothioamide
C1=CC(=C(C(=C1)Cl)Cl)C(=S)N
This item is not returnable. View return policy
84863-83-2
C7H5Cl2NS
206.084
MFCD00221163
PRRMJTDPKZBCCQ-UHFFFAOYSA-N
2,3-dichlorothiobenzamide, 2,3-dichloro-thiobenzamide, 2,3-dichlorobenzothioamide, 2,3-dichlorobenzene-1-carbothioamide, amino 2,3-dichlorophenyl methane-1-thione, dichlorothiobenzamide, pubchem22984, acmc-20aoh4, benzenecarbothioamide, 2,3-dichloro
2799029
2,3-dichlorobenzenecarbothioamide
C1=CC(=C(C(=C1)Cl)Cl)C(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 126°C to 128°C
Quantity 250 mg
Formula Weight 206.09
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorothiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'