missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Dichloropyridine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H63733.03

Code : NEW

Additional Details : CAS Number : 184416-84-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430579

  • £22.90 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

2,3-Dichloropyridine-4-carboxylic acid
192
2,3-Dichloroisonicotinic acid
184416-84-0
MFCD01861977

Specifications

2,3-Dichloropyridine-4-carboxylic acid
1g
Solid
192
2,3-Dichloroisonicotinic acid
203° to 207°C
184416-84-0
97%
MFCD01861977
Videos
SDS