missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzylamine, 98%, ACROS Organics™

Brand:  Acros Organics 365510010

Code : 29

Additional Details : CAS Number : 39226-95-4 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10374112

  • £12.80 / 1g


Estimated Shipment
25-06-2018
Add to basket


Specifications

Chemical Identifiers

2, 3-Dichlorobenzylamine
97.5
≥97.5%
C7H7Cl2N
176.05
39226-95-4
100.0
98%
Cl2C6H3CH2NH2
MFCD00047927

Specifications

2, 3-Dichlorobenzylamine
1g
97.5
1.3210g/mL
Glass bottle
Liquid
98%
Cl2C6H3CH2NH2
MFCD00047927
Clear colorless to light yellow
39226-95-4
100.0
Authentic
1.5820 to 1.5840 (20°C, 589nm)
≥97.5%
C7H7Cl2N
176.05
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,3-Dichlorobenzylamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS