missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzaldehyde, 99%, ACROS Organics™

Brand:  Acros Organics 232675000

Code : 29

Additional Details : CAS Number : 6334-18-5 Weight : 0.55000kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10760965

  • £543.40 / 500g


Estimated Shipment
21-06-2018
Add to basket


Specifications

Chemical Identifiers

2, 3-Dichlorobenzaldehyde
98.5
99%
C7H4Cl2O
175.01
6334-18-5
100.0
99%
Cl2C6H3CHO
MFCD00010127

Specifications

2, 3-Dichlorobenzaldehyde
144.0°C (30.0 mmHg)
60.0° to 64.0°C
6334-18-5
100.0
Authentic
Crystalline Powder
99%
Cl2C6H3CHO
MFCD00010127
135°C
White
500g
98.5
135g/mL
Glass bottle
99%
C7H4Cl2O
175.01
07, III, 878
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,3-Dichlorobenzaldehyde

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS