missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-6-fluorobenzylamine, 97%, Thermo Scientific™

Catalog Number 15403467
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
886501-27-5
C7H6Cl2FN
194.03
MFCD06660174
DQKOSZMSZGNUDK-UHFFFAOYSA-N
2,3-dichloro-6-fluorobenzylamine, 2,3-dichloro-6-fluorophenyl methanamine, 2,3-dichloro-6-fluorophenyl methylamine, 1-2,3-dichloro-6-fluorophenyl methanamine
17750750
(2,3-dichloro-6-fluorophenyl)methanamine
C1=CC(=C(C(=C1F)CN)Cl)Cl
This item is not returnable. View return policy
886501-27-5
C7H6Cl2FN
194.03
MFCD06660174
DQKOSZMSZGNUDK-UHFFFAOYSA-N
2,3-dichloro-6-fluorobenzylamine, 2,3-dichloro-6-fluorophenyl methanamine, 2,3-dichloro-6-fluorophenyl methylamine, 1-2,3-dichloro-6-fluorophenyl methanamine
17750750
(2,3-dichloro-6-fluorophenyl)methanamine
C1=CC(=C(C(=C1F)CN)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 194.04
Percent Purity 97%
Chemical Name or Material 2,3-Dichloro-6-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'