missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Dichloro-5-fluoropyridine, 98%

Brand:  Alfa Aesar™ H64373.03

UNSPSC : 12352101

Code : NEW

Additional Details : CAS Number : 185985-40-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15421549

  • £43.14 / 1g


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications

Description

Applications
2,3-Dichloro-5-fluoropyridine is a reagent that is used in the synthesis of 3-Chloro-5-fluoro Clodinafop Propargyl (C368645), which is an impurity of Clodinafop Propargyl.

Solubility
Slightly soluble in chloroform, methanol and water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

2,3-Dichloro-5-fluoropyridine
165.98
13831996
185985-40-4
MFCD08669666

Specifications

2,3-Dichloro-5-fluoropyridine
185985-40-4
98%
MFCD08669666
1g
Solid
165.98
13831996
Documentation

Technical Documents