missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propene, 98%, Thermo Scientific Chemicals

Catalog Number p-106023
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
78-88-6
C3H4Cl2
110.97
MFCD00000943
FALCMQXTWHPRIH-UHFFFAOYSA-N
2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41
6565
2,3-dichloroprop-1-ene
C=C(CCl)Cl
This item is not returnable. View return policy
78-88-6
C3H4Cl2
110.97
MFCD00000943
FALCMQXTWHPRIH-UHFFFAOYSA-N
2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41
6565
2,3-dichloroprop-1-ene
C=C(CCl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Brown-Pink to Colorless
Density 1.2000g/mL
Boiling Point 94°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(Cl)CH2Cl
Refractive Index 1.4601 to 1.4621
Quantity 100 mL
Beilstein 01, 199
Fieser 02,120; 08,158; 14,129
Specific Gravity 1.2
Formula Weight 110.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3-Dichloro-1-propene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dichloro-1-propene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'