missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-5-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Catalog Number 15417948
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
79623-38-4
C6H2Br2F3N
304.89
MFCD10687233
BIDFUURRDIAINQ-UHFFFAOYSA-N
26985667
2,3-dibromo-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CC(Br)=C(Br)N=C1
This item is not returnable. View return policy
79623-38-4
C6H2Br2F3N
304.89
MFCD10687233
BIDFUURRDIAINQ-UHFFFAOYSA-N
26985667
2,3-dibromo-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CC(Br)=C(Br)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 2,3-dibromo-5-trifluoromethyl pyridine, pyridine, 2,3-dibromo-5-trifluoromethyl, pyridine,2,3-dibromo-5-trifluoromethyl, pubchem15128, acmc-209phs, 2, 3-dibromo-5-trifluoromethyl pyridine
Formula Weight 304.89
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,3-Dibromo-5-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'