missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dibromo-3-phenylpropiophenone, 98%, ACROS Organics™
£9.44 - £23.21
Chemical Identifiers
CAS | 611-91-6 |
---|---|
Molecular Formula | C15H12Br2O |
Molecular Weight (g/mol) | 368.07 |
InChI Key | LYAGBKGGYRLVTR-UHFFFAOYSA-N |
Synonym | chalcone dibromide, 2,3-dibromo-1,3-diphenyl-1-propanone, benzalacetophenone dibromide, 2,3-dibromo-3-phenylpropiophenone, 1-propanone, 2,3-dibromo-1,3-diphenyl, propiophenone, 2,3-dibromo-3-phenyl, 1-propanone,2,3-dibromo-1,3-diphenyl, dibromodiphenylpropanone, benzal acetophenone dibromide, propiophenone,3-dibromo-3-phenyl |
PubChem CID | 95342 |
IUPAC Name | 2,3-dibromo-1,3-diphenylpropan-1-one |
SMILES | C1=CC=C(C=C1)C(C(C(=O)C2=CC=CC=C2)Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10016332
|
Acros Organics
375080010 |
1g | Glass bottle |
£9.44
1g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10641895
|
Acros Organics
375080050 |
5g | Glass bottle |
£23.21
5g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
611-91-6 | |
368.07 | |
chalcone dibromide, 2,3-dibromo-1,3-diphenyl-1-propanone, benzalacetophenone dibromide, 2,3-dibromo-3-phenylpropiophenone, 1-propanone, 2,3-dibromo-1,3-diphenyl, propiophenone, 2,3-dibromo-3-phenyl, 1-propanone,2,3-dibromo-1,3-diphenyl, dibromodiphenylpropanone, benzal acetophenone dibromide, propiophenone,3-dibromo-3-phenyl | |
2,3-dibromo-1,3-diphenylpropan-1-one |
C15H12Br2O | |
LYAGBKGGYRLVTR-UHFFFAOYSA-N | |
95342 | |
C1=CC=C(C=C1)C(C(C(=O)C2=CC=CC=C2)Br)Br |
Specifications
2, 3-Dibromo-3-phenylpropiophenone | |
156°C | |
611-91-6 | |
C15H12Br2O | |
chalcone dibromide, 2,3-dibromo-1,3-diphenyl-1-propanone, benzalacetophenone dibromide, 2,3-dibromo-3-phenylpropiophenone, 1-propanone, 2,3-dibromo-1,3-diphenyl, propiophenone, 2,3-dibromo-3-phenyl, 1-propanone,2,3-dibromo-1,3-diphenyl, dibromodiphenylpropanone, benzal acetophenone dibromide, propiophenone,3-dibromo-3-phenyl | |
LYAGBKGGYRLVTR-UHFFFAOYSA-N | |
2,3-dibromo-1,3-diphenylpropan-1-one | |
95342 | |
98% |
Glass bottle | |
1g | |
98.5% min. (HPLC) | |
C6H5CH(Br)CH(Br)C(O)C6H5 | |
Solubility in water: negligible | |
C1=CC=C(C=C1)C(C(C(=O)C2=CC=CC=C2)Br)Br | |
368.07 | |
368.07 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2,3-Dibromo-3-phenylpropiophenone
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.