missing translation for 'onlineSavingsMsg'
Learn More

2,3-Cyclopentenopyridine, 99+%, ACROS Organics™

Brand:  Acros Organics 188880050

Code : 29

Additional Details : CAS Number : 533-37-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10403530

  • £26.60 / 5g


Estimated Shipment
13-07-2018
Add to basket


Specifications

Chemical Identifiers

2, 3-Cyclopentenopyridine
99.0
99+%
119.17
533-37-9
100.0
C8H9N
6, 7-Dihydro-5H-cyclopenta[b]pyridine

Specifications

2, 3-Cyclopentenopyridine
67°C
5g
99.0
Glass bottle
99+%
C8H9N
119.17
1.0100g/mL
108.0° to 110.0°C (66.0 mbar)
533-37-9
100.0
99+%
1.5428 to 1.5448
1.01
6, 7-Dihydro-5H-cyclopenta[b]pyridine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,3-Cyclopentenopyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS