missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Cyclohexen-1-yl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62832.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439298

  • £125.08 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Cyclohexen-1-yl)thiazolidine
169.29
MFCD16685201

Specifications

2-(3-Cyclohexen-1-yl)thiazolidine
97%
MFCD16685201
1g
Liquid
169.29
Colorless
Videos
SDS