missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Chlorophenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62331.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15437296

  • £331.78 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Chlorophenyl)thiazolidine-4-carboxylic acid
243.71
MFCD08457608

Specifications

2-(3-Chlorophenyl)thiazolidine-4-carboxylic acid
Solid
243.71
5g
97%
MFCD08457608
Videos
SDS