missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chlorophenyl)benzimidazole, 95%, Thermo Scientific™

Catalog Number 15471349
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
22868-35-5
C13H9ClN2
228.68
MFCD01438282
BGYHTIIVIGGCIF-UHFFFAOYSA-N
767074
ClC1=CC=CC(=C1)C1=NC2=CC=CC=C2N1
This item is not returnable. View return policy
22868-35-5
C13H9ClN2
228.68
MFCD01438282
BGYHTIIVIGGCIF-UHFFFAOYSA-N
767074
ClC1=CC=CC(=C1)C1=NC2=CC=CC=C2N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 2-3-chlorophenyl-1h-benzimidazole, 2-3-chlorophenyl benzimidazole, 2-3-chlorophenyl-1h-1,3-benzodiazole, 2-3-chlorophenyl-1h-benzo d imidazole, 2-3-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-3-chlorophenyl, cbmicro_048649, 1h-benzimidazole,2-3-chlorophenyl
IUPAC Name 2-(3-chlorophenyl)-1H-benzimidazole
Formula Weight 228.68
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(3-Chlorophenyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'