Alfa Aesar™ 2-(3-Chlorophenyl)benzimidazole, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64245.06

UNSPSC: 12352005

Code: NEW

Additional Details:
Additional Details: CAS Number: 22868-35-5 Weight: 0.00500kg


Quantity 5g

Product Code. 15461349

Quantity Price
1 £ 535.3 / 5g
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-(3-Chlorophenyl)benzimidazole
CAS 22868-35-5
Formula Weight 228.68
MDL Number MFCD01438282

Specification

Formula Weight 228.68
Quantity 5g
MDL Number MFCD01438282
CAS 22868-35-5
Chemical Name or Material 2-(3-Chlorophenyl)benzimidazole
Physical Form Solid
Assay 95%