missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Chlorophenyl)-5-methylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64198.06

Code : NEW

Additional Details : CAS Number : 392720-65-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15481299

  • £649.78 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Chlorophenyl)-5-methylbenzimidazole
392720-65-9
242.71
MFCD01836514

Specifications

2-(3-Chlorophenyl)-5-methylbenzimidazole
392720-65-9
95%
MFCD01836514
5g
Solid
242.71
Videos
SDS