missing translation for 'onlineSavingsMsg'
Learn More

(2-(3-Chlorophenyl)-1,3-thiazol-4-yl)methanamine hydrochloride monohydrate, 97%, Thermo Scientific™

Catalog Number p-3075125
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
690632-12-3
C10H10Cl2N2S
261.164
MFCD06738410
DBEFCUHDXFEPSJ-UHFFFAOYSA-N
2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl thiazol-4-yl methylamine hydrochloride, c-2-3-chloro-phenyl-thiazol-4-yl-methylamine hydrochloride, 2-3-chlorophenyl-1,3-thiazol-4-yl methane amine hydrochloride 1-hydrate, 4-aminomethyl-2-3-chlorophenyl thiazole hcl, 2-3-chlorophenyl thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl-4-thiazolyl methanamine hydrochloride, 2-3-chloro-phenyl-thiazol-4-ylmethyllamine hcl, 2-3-chlorophenyl-1,3-thiazol-4-yl methylamine, chloride, 1-2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride
24229776
[2-(3-chlorophenyl)-1,3-thiazol-4-yl]methanamine;hydrochloride
C1=CC(=CC(=C1)Cl)C2=NC(=CS2)CN.Cl
This item is not returnable. View return policy
690632-12-3
C10H10Cl2N2S
261.164
MFCD06738410
DBEFCUHDXFEPSJ-UHFFFAOYSA-N
2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl thiazol-4-yl methylamine hydrochloride, c-2-3-chloro-phenyl-thiazol-4-yl-methylamine hydrochloride, 2-3-chlorophenyl-1,3-thiazol-4-yl methane amine hydrochloride 1-hydrate, 4-aminomethyl-2-3-chlorophenyl thiazole hcl, 2-3-chlorophenyl thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl-4-thiazolyl methanamine hydrochloride, 2-3-chloro-phenyl-thiazol-4-ylmethyllamine hcl, 2-3-chlorophenyl-1,3-thiazol-4-yl methylamine, chloride, 1-2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride
24229776
[2-(3-chlorophenyl)-1,3-thiazol-4-yl]methanamine;hydrochloride
C1=CC(=CC(=C1)Cl)C2=NC(=CS2)CN.Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 279.2
Percent Purity 97%
Chemical Name or Material [2-(3-Chlorophenyl)-1, 3-thiazol-4-yl]methanamine hydrochloride monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'