missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-(3-Chlorophenyl)-1,3-thiazol-4-yl)methanamine hydrochloride monohydrate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 279.2 |
Percent Purity | 97% |
Chemical Name or Material | [2-(3-Chlorophenyl)-1, 3-thiazol-4-yl]methanamine hydrochloride monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'