missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Chloromethyl)phenyl]pyrimidine, 97%, Thermo Scientific™

Catalog Number p-3522442
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
898289-51-5
C11H9ClN2
204.657
MFCD09064978
AUWJTKGKFPDHDU-UHFFFAOYSA-N
2-3-chloromethyl phenyl pyrimidine, 3-pyrimidin-2-ylbenzyl chloride, pyrimidine, 2-3-chloromethyl phenyl, pyrimidine, 2-3-chloromethyl phenyl-9ci
24229567
2-[3-(chloromethyl)phenyl]pyrimidine
C1=CC(=CC(=C1)CCl)C2=NC=CC=N2
This item is not returnable. View return policy
898289-51-5
C11H9ClN2
204.657
MFCD09064978
AUWJTKGKFPDHDU-UHFFFAOYSA-N
2-3-chloromethyl phenyl pyrimidine, 3-pyrimidin-2-ylbenzyl chloride, pyrimidine, 2-3-chloromethyl phenyl, pyrimidine, 2-3-chloromethyl phenyl-9ci
24229567
2-[3-(chloromethyl)phenyl]pyrimidine
C1=CC(=CC(=C1)CCl)C2=NC=CC=N2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.66
Percent Purity 97%
Chemical Name or Material 2-[3-(Chloromethyl)phenyl]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'