missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Chloromethyl)phenyl]-1,3-thiazole, 97%, Maybridge

Brand:  Maybridge CC36524DA

Code : 31

Additional Details : CAS Number : 906352-60-1 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10538823

  • £83.00 / 1g


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Specifications

Chemical Identifiers

2-[3-(Chloromethyl)phenyl]-1, 3-thiazole
96.5
97%
C10H8ClNS
MFCD09025829
906352-60-1
100.0
97%
209.7

Specifications

2-[3-(Chloromethyl)phenyl]-1, 3-thiazole
96.5
Amber Glass Bottle
97%
C10H8ClNS
MFCD09025829
906352-60-1
100.0
97%
1g
209.7
Videos
SDS