missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Butynyloxy)tetrahydro-2H-pyran, 97%, ACROS Organics™

Brand:  Acros Organics 428210250

Code : 29

Additional Details : CAS Number : 40365-61-5 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10547600

  • £164.20 / 25g


Estimated Shipment
14-03-2018
Add to basket


Specifications

Chemical Identifiers

2-(3-Butynyloxy)tetrahydro-2H-pyran
96.0
97%
154.21
40365-61-5
100.0
C9H14O2
MFCD00012352

Specifications

2-(3-Butynyloxy)tetrahydro-2H-pyran
96.0
97%
C9H14O2
154.21
0.9840g/mL
25g
40365-61-5
100.0
97%
Glass bottle
MFCD00012352
92.0° to 95.0°C (18.0 mmHg)
73°C
Videos
SDS