missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione, 99%, Thermo Scientific Chemicals

Catalog Number p-7021751
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
431-03-8
C4H6O2
86.09
MFCD00008756
QSJXEFYPDANLFS-UHFFFAOYSA-N
2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
650
CHEBI:16583
butane-2,3-dione
CC(=O)C(=O)C
This item is not returnable. View return policy
431-03-8
C4H6O2
86.09
MFCD00008756
QSJXEFYPDANLFS-UHFFFAOYSA-N
2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
650
CHEBI:16583
butane-2,3-dione
CC(=O)C(=O)C

2,3-Butanedione is used to form triazine and pteridine ring systems by cyclocondensation with amines. It is also used to inactivate aminopeptidase-N and acts as a precursor to alpha-diones. It finds use in alcoholic beverages, foods, to modify arginyl residues in proteins and to give a buttery flavor to microwave popcorns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Butanedione is used to form triazine and pteridine ring systems by cyclocondensation with amines. It is also used to inactivate aminopeptidase-N and acts as a precursor to alpha-diones. It finds use in alcoholic beverages, foods, to modify arginyl residues in proteins and to give a buttery flavor to microwave popcorns.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with acids, strong bases, metals, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -3°C
Density 0.983
Boiling Point 87°C to 88°C
Flash Point 10°C (50°F)
Odor Sweet
Refractive Index 1.394
Quantity 10 g
UN Number UN2346
Beilstein 605398
Merck Index 14,2966
Solubility Information Miscible with water.
Formula Weight 86.09
Percent Purity 99%
Chemical Name or Material 2,3-Butanedione
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'