Learn More
Alfa Aesar™ 2-(3-Bromophenyl)thiazolidine, 97%
Brand: Alfa Aesar™ H62658.03
Code : NEW
Additional Details : CAS Number : 67086-84-4 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2-(3-Bromophenyl)thiazolidine | |
67086-84-4 | |
244.15 | |
MFCD00222816 |
Specifications
2-(3-Bromophenyl)thiazolidine | |
1g | |
97% | |
MFCD00222816 |
67086-84-4 | |
Solid | |
244.15 |