missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Bromophenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62875.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457946

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

2-(3-Bromophenyl)thiazolidine-4-carboxylic acid
288.16
MFCD00176218

Specifications

2-(3-Bromophenyl)thiazolidine-4-carboxylic acid
Solid
288.16
1g
97%
MFCD00176218
Videos
SDS